Ethyl-2-bromo-5-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromo-5-nitrobenzoate

075816-1G 075816-5G 075816-10G
102543-146EA 427.69 USD
102543-146 102543-148 102543-150
Ethyl-2-bromo-5-nitrobenzoate ≥95%
Ethyl-2-bromo-5-nitrobenzoate

Hazard: Irritant

Formula: C₉H₈BrNO₄
MW: 274.0700 g/mol
MDL Number: MFCD12025203
CAS Number: 208176-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR