(R)-(+)-Tetrahydro-2-furoic acid ≥95%

Supplier: Matrix Scientific

075815-5G 075815-25G
102543-142EA 106.15 USD
102543-142 102833-876
(R)-(+)-Tetrahydro-2-furoic acid ≥95%
(R)-(+)-Tetrahydro-2-furoic acid

Hazard: Irritant

Formula: C₅H₈O₃
MDL Number: MFCD00211271
CAS Number: 87392-05-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR