2-(4-Bromo-1H-indol-3-yl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromoindole-3-acetic Acid

075814-250MG 075814-1G 075814-5G
102543-134EA 355.38 USD
102543-134 102543-136 102543-138
2-(4-Bromo-1H-indol-3-yl)acetic acid ≥95%
2-(4-Bromo-1H-indol-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₈BrNO₂
MW: 254.0800 g/mol
MDL Number: MFCD09751717
CAS Number: 89245-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR