4-Bromo-3-nitroaniline ≥95%

Supplier: Matrix Scientific

075813-1G 075813-5G 075813-10G
102543-128EA 143.08 USD
102543-128 102543-130 102543-132
4-Bromo-3-nitroaniline ≥95%
4-Bromo-3-nitroaniline

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MDL Number: MFCD01320678
CAS Number: 53324-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR