Meropenem ≥95%
Supplier: Matrix Scientific
Synonyms:
(4R,5S,6S)-3-[[(3S,5S)-5-[(dimethylamino)carbonyl]-3-pyrrolidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, ICI 194660, Merrem® I.V., SM-7338, (4R,5S,6S)-3-(((3S,5S)-5-(Dimethylcarbamoyl)pyrrolidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid
075811-1G
075811-5G
102543-118EA
463.08
USD
102543-118
102543-120
Meropenem ≥95%
Meropenem
Hazard code(s): Irritant
Formula:
C₁₇H₂₅N₃O₅S MW: 383.5 g/mol |
MDL Number:
MFCD00864966 CAS Number: 96036-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...