β-D-Ribofuranose-1,2,3,5-tetraacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Tetra-O-acetyl-β-D-ribofuranose, (2S,3R,4R,5R)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate

075810-5G 075810-10G 075810-100G
102543-112EA 15.38 USD
102543-112 102543-114 102543-116
β-D-Ribofuranose-1,2,3,5-tetraacetate ≥95%
beta-D-Ribofuranose-1,2,3,5-tetraacetate

Hazard: Irritant

Formula: C₁₃H₁₈O₉
Melting Pt: 80…83 °C
MDL Number: MFCD00005358
CAS Number: 13035-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR