3-Hydroxy-4-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

075804-1G 075804-5G 075804-10G
102543-086EA 178.46 USD
102543-086 102543-088 102543-090
3-Hydroxy-4-methylbenzonitrile ≥95%
3-Hydroxy-4-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₇NO
MW: 133.1500 g/mol
MDL Number: MFCD08361769
CAS Number: 3816-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR