Thiazolidine-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1,3-Thiazolidine-2-carboxylic acid, 2-Carboxy-1,3-thiazolidine, 1,3-Thiazolane-2-carboxylic acid
075803-1G
075803-5G
075803-10G
102543-080EA
55.38
USD
102543-080
102543-082
102543-084
Thiazolidine-2-carboxylic acid ≥95%
Thiazolidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₄H₇NO₂S MW: 133.169 g/mol |
MDL Number:
MFCD00068005 CAS Number: 65126-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...