tert-Butyl-4-amino-3-methylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-amino-3-methylbenzoate

075802-1G 075802-5G
102543-076EA 533.85 USD
102543-076 102543-078
tert-Butyl-4-amino-3-methylbenzoate ≥95%
tert-Butyl-4-amino-3-methylbenzoate

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MW: 207.2689 g/mol
MDL Number: MFCD08703398
CAS Number: 934481-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR