Ethyl carbamimidate hydrochloride ≥95%

Supplier: Matrix Scientific

075801-1G 075801-5G 075801-25G
102543-070EA 106.15 USD
102543-070 102543-072 102833-874
Ethyl carbamimidate hydrochloride ≥95%
Ethyl carbamimidate hydrochloride

Hazard: Irritant

Formula: C₃H₉ClN₂O
MW: 124.5694 g/mol
MDL Number: MFCD07438010
CAS Number: 31407-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR