(-)-Dimethyl-L-malate ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(-)-Dimethyl Malate, (S)-Dimethyl 2-hydroxysuccinate, Dimethyl (S)-(-)-malate

075799-10G 075799-100G
102543-062EA 64.62 USD
102543-062 102543-064
(-)-Dimethyl-L-malate ≥95%
(-)-Dimethyl-L-malate

Hazard: Irritant

Formula: C₆H₁₀O₅
MW: 162.1406 g/mol
MDL Number: MFCD00066215
CAS Number: 617-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR