3-Bromo-2-chloro-4-methylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-2-chloro-4-picoline
075796-1G
075796-5G
102543-048EA
269.23
USD
102543-048
102543-050
3-Bromo-2-chloro-4-methylpyridine ≥95%
3-Bromo-2-chloro-4-methylpyridine
Hazard: Irritant
Formula:
C₆H₅BrClN MW: 206.47 g/mol |
MDL Number:
MFCD04112511 CAS Number: 55404-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...