Picolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid

075795-100G 075795-500G
102543-046EA 56.92 USD
102543-046 102551-996
Picolinic acid ≥95%
Picolinic acid

Hazard: Irritant

Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 293
Melting Pt: 135…139 °C
MDL Number: MFCD00006293
CAS Number: 98-98-6
Merck Index: 12,07557

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR