(R)-1,4-Diazabicyclo[4.3.0]nonane ≥95%

Supplier: Matrix Scientific

075794-1G 075794-5G
102543-040EA 783.08 USD
102543-040 102543-042
(R)-1,4-Diazabicyclo[4.3.0]nonane ≥95%
(R)-1,4-Diazabicyclo[4.3.0]nonane

Hazard: Irritant

Formula: C₇H₁₄N₂
MDL Number: MFCD09752820
CAS Number: 96193-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR