2-Methyl-1-pyrroline ≥95%

Supplier: Matrix Scientific

075792-5G 075792-10G 075792-100G
102543-032EA 143.08 USD
102543-032 102543-034 102543-036
2-Methyl-1-pyrroline ≥95%
2-Methyl-1-pyrroline

Hazard: Flammable/Corrosive

Formula: C₅H₉N
MW: 83.13 g/mol
Boiling Pt: 104…105 °C
Density: 0.878
Flash Pt: 10 °C
MDL Number: MFCD00128806
CAS Number: 872-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR