1-(5-Bromo-2,4-difluorophenyl)ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2,4-difluoroacetophenone, 5'-Bromo-2',4'-difluoroacetophenone

075791-1G 075791-5G
102543-026EA 250.77 USD
102543-026 102543-028
1-(5-Bromo-2,4-difluorophenyl)ethanone ≥95%
1-(5-Bromo-2,4-difluorophenyl)ethanone

Hazard: Irritant

Formula: C₈H₅BrF₂O
MW: 235.0255 g/mol
MDL Number: MFCD13190613
CAS Number: 864773-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR