2-Nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

075783-10G 075783-100G
102542-996EA 498.46 USD
102542-996 102542-998
2-Nitrobenzoic acid ≥95%
2-Nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₅NO₄
Melting Pt: 146-148°C
MDL Number: MFCD00007137
CAS Number: 552-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR