4-Methylquinoline ≥95%
Supplier: Matrix Scientific
075780-5G
075780-10G
075780-100G
102542-982EA
816.92
USD
102542-982
102542-984
102542-986
4-Methylquinoline ≥95%
4-Methylquinoline
Hazard: Irritant
Formula:
C₁₀H₉N Boiling Pt: 260…263 °C Melting Pt: 9…10 °C Density: 1.083 Flash Pt: >110 °C |
MDL Number:
MFCD00006784 CAS Number: 491-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...