2-Bromo-4-pyridineboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2-Bromo-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Bromopyridine-4-boronic acid pinacol ester, 2-Bromo-4-pyridylboronic acid Pinacol ester

075779-1G 075779-5G
102542-978EA 116.92 USD
102542-978 102542-980
2-Bromo-4-pyridineboronic acid pinacol ester ≥95%
2-Bromo-4-pyridineboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₁H₁₅BBrNO₂
Melting Pt: 80-83°C
MDL Number: MFCD06798252
CAS Number: 458532-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR