Boc-Glu-OtBu ≥95%

Supplier: Matrix Scientific

Synonyms: N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-tert-Butyl Ester, N-Boc-L-glutamic Acid 1-tert-Butyl Ester, 1-tert-Butyl N-Boc-L-glutamate, Boc-Glu-OtBu

075776-5G 075776-25G
102542-964EA 44.62 USD
102542-964 102833-872
Boc-Glu-OtBu ≥95%
Boc-Glu-OtBu

Hazard: Irritant

Formula: C₁₄H₂₅NO₆
MW: 303.36 g/mol
MDL Number: MFCD00038273
CAS Number: 24277-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR