(R)-3-Hydroxymethylmorpholine ≥95%

Supplier: Matrix Scientific

075775-250MG 075775-1G 075775-5G
102542-956EA 1458.46 USD
102542-956 102542-958 102542-960
(R)-3-Hydroxymethylmorpholine ≥95%
(R)-3-Hydroxymethylmorpholine

Hazard: Irritant

Formula: C₅H₁₁NO₂
MW: 117.1500 g/mol
MDL Number: MFCD06799479
CAS Number: 211053-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR