Ethyl-2-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-Piperidinecarboxylate, Ethyl pipecolinate

075772-5G 075772-100G 075772-25G
102542-942EA 64.62 USD
102542-942 102542-946 102551-994
Ethyl-2-piperidinecarboxylate ≥95%
Ethyl 2-piperidinecarboxylate

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD00005980
CAS Number: 15862-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR