DL-α-Phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: H-DL-Phe-OH, DL-Phenylalanine, 3-Phenyl-DL-alanine

075771-10G 075771-100G
102542-938EA 36.92 USD
102542-938 102542-940
DL-α-Phenylalanine ≥95%
DL-α-Phenylalanine

Hazard: Irritant/Refrigerate

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313
Melting Pt: 173
MDL Number: MFCD00064225
CAS Number: 150-30-1
Merck Index: 13,07355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR