(R)-Ethyl-2-amino-3-hydroxypropanoate hydrochloride ≥95%

Supplier: Matrix Scientific

075768-1G 075768-5G 075768-10G
102542-928EA 36.92 USD
102542-928 102542-930 102542-932
(R)-Ethyl-2-amino-3-hydroxypropanoate hydrochloride ≥95%
(R)-Ethyl-2-amino-3-hydroxypropanoate hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO₃
MDL Number: MFCD00191020
CAS Number: 104055-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR