Ribavirin ≥95%
Supplier: Matrix Scientific
Synonyms:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-1,2,4-triazole-3-carboxamide
075767-10G
075767-100G
102542-924EA
53.85
USD
102542-924
102542-926
Ribavirin ≥95%
Ribavirin
Hazard: Irritant
Formula:
C8H12N4O5 MW: 244.2047 g/mol |
MDL Number:
MFCD00058564 CAS Number: 36791-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...