3-Fluoro-2-methoxybenzyl alcohol ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-2-methoxybenzenemethanol
075766-1G
075766-5G
102542-920EA
396.92
USD
102542-920
102542-922
3-Fluoro-2-methoxybenzyl alcohol ≥95%
3-Fluoro-2-methoxybenzyl alcohol
Hazard: Irritant
Formula:
C₈H₉FO₂ MW: 156.16 g/mol |
MDL Number:
MFCD09025369 CAS Number: 303043-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...