(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: L-Cysteine ethyl ester hydrochloride, H-Cys-Oet.HCl, H-Cys-OEt·HCl

075765-10G 075765-100G
102542-914EA 47.69 USD
102542-914 102542-916
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥95%
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride

Hazard: Irritant

Formula: C₅H₁₁NO₂S·HCl
MW: 185.666 g/mol
MDL Number: MFCD00012631
CAS Number: 868-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR