O-Benzylhydroxylamine hydrochloride ≥95%

Supplier: Matrix Scientific

075763-100G 075763-25G
102542-908EA 127.69 USD
102542-908 102833-870
O-Benzylhydroxylamine hydrochloride ≥95%
O-Benzylhydroxylamine hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNO
MW: 159.62 g/mol
MDL Number: MFCD00012952
CAS Number: 2687-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR