Vildagliptin ≥95%

Supplier: Matrix Scientific

Synonyms: (2S)-1-((3-Hydroxyadamantan-1-yl)glycyl)pyrrolidine-2-carbonitrile

075762-1G 075762-5G
102542-902EA 384.62 USD
102542-902 102542-904
Vildagliptin ≥95%
Vildagliptin

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₂
MW: 303.3993 g/mol
MDL Number: MFCD08275142
CAS Number: 274901-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR