4-Chloro-o-cresol ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Chloro-4-hydroxy-3-methylbenzene, 4-Chloro-2-methylphenol
075761-10G
075761-100G
102542-898EA
107.69
USD
102542-898
102542-900
4-Chloro-o-cresol ≥95%
4-Chloro-o-cresol
Hazard: Irritant
Formula:
C₇H₇ClO |
MDL Number:
MFCD00002321 CAS Number: 1570-64-5 UN: 3437 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...