4-Bromobenzylamine ≥95%
Supplier: Matrix Scientific
075758-100G
075758-25G
102542-886EA
264.62
USD
102542-886
102664-064
4-Bromobenzylamine ≥95%
4-Bromobenzylamine
Hazard: Corrosive/Refrigerate
Formula:
C₇H₈BrN MW: 186.05 g/mol Boiling Pt: 250 °C Melting Pt: 25 °C |
MDL Number:
MFCD00047931 CAS Number: 3959-07-7 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...