2-Methoxy-4-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

075757-250MG 075757-1G
102542-878EA 640 USD
102542-878 102542-880
2-Methoxy-4-methylbenzonitrile ≥95%
2-Methoxy-4-methylbenzonitrile

Hazard: Irritant

Formula: C₉H₉NO
MDL Number: MFCD11847156
CAS Number: 53078-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR