4-Bromo-N,N-dimethylpyridin-2-amine ≥95%

Supplier: Matrix Scientific

075756-250MG 075756-1G
102542-874EA 541.54 USD
102542-874 102542-876
4-Bromo-N,N-dimethylpyridin-2-amine ≥95%
4-Bromo-N,N-dimethylpyridin-2-amine

Hazard: Irritant

Formula: C₇H₉BrN₂
MDL Number: MFCD11869615
CAS Number: 946000-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR