5-Formylpicolinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Formylpyridine-2-carbonitrile

075755-1G 075755-5G
102542-870EA 515.38 USD
102542-870 102542-872
5-Formylpicolinonitrile ≥95%
5-Formylpicolinonitrile

Hazard: Irritant

Formula: C7H4N2O
MW: 132.1200 g/mol
MDL Number: MFCD12755972
CAS Number: 131747-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR