3-Chloro-5-fluorobenzaldehyde ≥95%

Supplier: Matrix Scientific

075754-1G 075754-5G 075754-10G
102542-862EA 143.08 USD
102542-862 102542-864 102542-866
3-Chloro-5-fluorobenzaldehyde ≥95%
3-Chloro-5-fluorobenzaldehyde

Hazard: Irritant

Formula: C₇H₄ClFO
MW: 158.56 g/mol
MDL Number: MFCD02261734
CAS Number: 90390-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR