9-Bromo-10-(naphthalen-1-yl)anthracene ≥95%

Supplier: Matrix Scientific

075748-1G 075748-5G
102542-836EA 533.85 USD
102542-836 102542-838
9-Bromo-10-(naphthalen-1-yl)anthracene ≥95%
9-Bromo-10-(naphthalen-1-yl)anthracene

Hazard: Irritant

Formula: C₂₄H₁₅Br
MDL Number: MFCD11046571
CAS Number: 400607-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR