(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid ≥95%

Supplier: Matrix Scientific

075747-1G 075747-5G 075747-10G
102542-830EA 233.85 USD
102542-830 102542-832 102542-834
(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid ≥95%
(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid

Hazard: Irritant

Formula: C₈H₉BClFO₃
MW: 218.4177 g/mol
MDL Number: MFCD15144702
CAS Number: 909122-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR