Cbz-D-Ser-OH ≥95%
Supplier: Matrix Scientific
075746-1G
075746-5G
075746-10G
102542-824EA
55.38
USD
102542-824
102542-826
102542-828
Cbz-D-Ser-OH ≥95%
Cbz-D-Ser-OH
Hazard: Irritant
Formula:
C₁₁H₁₃NO₅ |
MDL Number:
MFCD00063144 CAS Number: 6081-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...