1-(2-Chloroethyl)piperidinium chloride ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2-Chloroethyl)-piperidine hydrochloride, 1-(2-Chloroethyl)piperidine hydrochloride

075742-10G 075742-100G
102542-808EA 36.92 USD
102542-808 102542-810
1-(2-Chloroethyl)piperidinium chloride ≥95%
1-(2-Chloroethyl)piperidinium chloride

Hazard: Irritant

Formula: C₇H₁₅Cl₂N
MW: 184.11 g/mol
MDL Number: MFCD00012837
CAS Number: 2008-75-5
UN: 2928
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR