1-(2-Bromoethoxy)-2-ethoxybenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Ethoxyphenoxy)ethyl bromide

075740-5G 075740-10G
102542-798EA 107.69 USD
102542-798 102542-800
1-(2-Bromoethoxy)-2-ethoxybenzene ≥95%
1-(2-Bromoethoxy)-2-ethoxybenzene

Hazard: Irritant

Formula: C₁₀H₁₃BrO₂
MW: 245.12 g/mol
Boiling Pt: 142-145°C/11 Torr
MDL Number: MFCD02030483
CAS Number: 3259-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR