2-tert-Butyl-4-ethyl-3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(tert-Butyl) 4-ethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate

075738-1G 075738-5G 075738-10G
102542-788EA 143.08 USD
102542-788 102542-790 102542-792
2-tert-Butyl-4-ethyl-3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate ≥95%
2-tert-Butyl-4-ethyl-3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₂₁NO₄
MW: 267.3208 g/mol
MDL Number: MFCD02179394
CAS Number: 86770-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR