2-Chloro-5-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

075736-10G 075736-100G
102542-778EA 36.92 USD
102542-778 102542-780
2-Chloro-5-nitrobenzoic acid ≥95%
2-Chloro-5-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Melting Pt: 166-168°C
MDL Number: MFCD00007294
CAS Number: 2516-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR