4,5-Dimethyl-1,3-dioxol-2-one ≥95%

Supplier: Matrix Scientific

075732-10G 075732-100G
102542-762EA 32.31 USD
102542-762 102542-764
4,5-Dimethyl-1,3-dioxol-2-one ≥95%
4,5-Dimethyl-1,3-dioxol-2-one

Hazard: Irritant

Formula: C₅H₆O₃
MW: 114.0993 g/mol
MDL Number: MFCD00135139
CAS Number: 37830-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR