2,6-Dichlorobenzothiazole ≥95%

Supplier: Matrix Scientific

075731-1G 075731-5G 075731-10G
102542-756EA 90.77 USD
102542-756 102542-758 102542-760
2,6-Dichlorobenzothiazole ≥95%
2,6-Dichlorobenzothiazole

Hazard: Irritant

Formula: C₇H₃Cl₂NS
MDL Number: MFCD00044101
CAS Number: 3622-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR