Methyl-4-hydroxyquinoline-2-carboxylate ≥95%

Supplier: Matrix Scientific

075730-1G 075730-5G 075730-10G
102542-750EA 161.54 USD
102542-750 102542-752 102542-754
Methyl-4-hydroxyquinoline-2-carboxylate ≥95%
Methyl 4-hydroxyquinoline-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₉NO₃
MDL Number: MFCD08693316
CAS Number: 5965-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR