2,2'-Dibromobiphenyl (PBB No. 4) ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-2-(2-bromophenyl)benzene, 2,2'-Dibromobiphenyl

075727-1G 075727-5G 075727-25G
102542-734EA 30.77 USD
102542-734 102542-736 102833-868
2,2'-Dibromobiphenyl (PBB No. 4) ≥95%
2,2'-Dibromobiphenyl (PBB No. 4)

Hazard: Irritant

Formula: C₁₂H₈Br₂
MW: 312 g/mol
Melting Pt: 78…81 °C
MDL Number: MFCD00093707
CAS Number: 13029-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR