4-Chloro-1-methylpiperidine ≥95%
Supplier: Matrix Scientific
075722-10G
075722-100G
102542-710EA
33.85
USD
102542-710
102542-712
4-Chloro-1-methylpiperidine ≥95%
4-Chloro-1-methylpiperidine
Hazard: Irritant
Formula:
C₆H₁₂ClN MW: 133.6192 g/mol |
MDL Number:
MFCD00044489 CAS Number: 5570-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...