(S)-2-Aminobutanamide hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: H-Abu-NH2 · HCl, (2S)-2-Aminobutanamide hydrochloride, (S)-(+)-2-Aminobutanamide hydrochloride, H-Abu-NH2 HCl

075721-1G 075721-5G 075721-10G
102542-704EA 813.85 USD
102542-704 102542-706 102542-708
(S)-2-Aminobutanamide hydrochloride ≥95%
(S)-2-Aminobutanamide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂O
MW: 138.60 g/mol
MDL Number: MFCD00136565
CAS Number: 7682-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR