Methyl-5-bromo-2-hydroxy-3-methoxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-2-hydroxy-3-methoxybenzoate

075718-1G 075718-5G 075718-10G
102542-686EA 533.85 USD
102542-686 102542-688 102542-690
Methyl-5-bromo-2-hydroxy-3-methoxybenzoate ≥95%
Methyl-5-bromo-2-hydroxy-3-methoxybenzoate

Hazard: Irritant

Formula: C₉H₉BrO₄
MW: 261.0700 g/mol
MDL Number: MFCD06203682
CAS Number: 134419-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR