4-Fluoro-2-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

075717-1G 075717-5G 075717-10G
102542-680EA 110.77 USD
102542-680 102542-682 102542-684
4-Fluoro-2-nitrobenzonitrile ≥95%
4-Fluoro-2-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₃FN₂O₂
MDL Number: MFCD00277447
CAS Number: 80517-21-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR